missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenyl Ditelluride 98.0+%, TCI America™
$231.02 - $691.13
Chemical Identifiers
| CAS | 32294-60-3 |
|---|---|
| Molecular Formula | C12H10Te2 |
| Molecular Weight (g/mol) | 409.412 |
| MDL Number | MFCD00192106 |
| InChI Key | VRLFOXMNTSYGMX-UHFFFAOYSA-N |
| Synonym | diphenyl ditelluride, phenyl ditelluride, ditelluride, diphenyl, diphenylditelluride, ditelluride, diephenyl, phenylditellanyl benzene, diphenylditellane, diphenyl pertelluride, acmc-209hsa, 1,2-diphenylditellane # |
| PubChem CID | 100657 |
| IUPAC Name | (phenylditellanyl)benzene |
| SMILES | C1=CC=C(C=C1)[Te][Te]C2=CC=CC=C2 |
Chemical Identifiers
| 32294-60-3 | |
| 409.412 | |
| VRLFOXMNTSYGMX-UHFFFAOYSA-N | |
| 100657 | |
| C1=CC=C(C=C1)[Te][Te]C2=CC=CC=C2 |
| C12H10Te2 | |
| MFCD00192106 | |
| diphenyl ditelluride, phenyl ditelluride, ditelluride, diphenyl, diphenylditelluride, ditelluride, diephenyl, phenylditellanyl benzene, diphenylditellane, diphenyl pertelluride, acmc-209hsa, 1,2-diphenylditellane # | |
| (phenylditellanyl)benzene |
Specifications
| 32294-60-3 | |
| C12H10Te2 | |
| 1 g | |
| diphenyl ditelluride, phenyl ditelluride, ditelluride, diphenyl, diphenylditelluride, ditelluride, diephenyl, phenylditellanyl benzene, diphenylditellane, diphenyl pertelluride, acmc-209hsa, 1,2-diphenylditellane # | |
| C1=CC=C(C=C1)[Te][Te]C2=CC=CC=C2 | |
| 409.412 | |
| 409.41 | |
| Crystalline Powder |
| 65°C | |
| MFCD00192106 | |
| 3284 | |
| VRLFOXMNTSYGMX-UHFFFAOYSA-N | |
| (phenylditellanyl)benzene | |
| 100657 | |
| ≥98.0% (GC) | |
| Diphenyl Ditelluride |
Safety and Handling
TSCA : No