missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Ditelluride 98.0+%, TCI America™

Catalog No. D27181G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27181G 1 g
D27185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27181G Supplier TCI America Supplier No. D27181G

Chemical Identifiers

CAS 32294-60-3
Molecular Formula C12H10Te2
Molecular Weight (g/mol) 409.412
MDL Number MFCD00192106
InChI Key VRLFOXMNTSYGMX-UHFFFAOYSA-N
Synonym diphenyl ditelluride, phenyl ditelluride, ditelluride, diphenyl, diphenylditelluride, ditelluride, diephenyl, phenylditellanyl benzene, diphenylditellane, diphenyl pertelluride, acmc-209hsa, 1,2-diphenylditellane #
PubChem CID 100657
IUPAC Name (phenylditellanyl)benzene
SMILES C1=CC=C(C=C1)[Te][Te]C2=CC=CC=C2

Specifications

Melting Point 65°C
Quantity 1 g
UN Number 3284
Formula Weight 409.41
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyl Ditelluride