missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl ditelluride, 97%

Catalog No. AC301840010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC301840010 1 g Glass bottle
AC301840050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC301840010 Supplier Thermo Scientific Chemicals Supplier No. 301840010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 32294-60-3 | C12H10Te2 | 409.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32294-60-3
Molecular Formula C12H10Te2
Molecular Weight (g/mol) 409.27
MDL Number MFCD00192106
InChI Key VRLFOXMNTSYGMX-UHFFFAOYSA-N
Synonym diphenyl ditelluride, phenyl ditelluride, ditelluride, diphenyl, diphenylditelluride, ditelluride, diephenyl, phenylditellanyl benzene, diphenylditellane, diphenyl pertelluride, acmc-209hsa, 1,2-diphenylditellane #
PubChem CID 100657
IUPAC Name (phenylditellanyl)benzene
SMILES C1=CC=C(C=C1)[Te][Te]C2=CC=CC=C2

Specifications

Melting Point 64°C to 67°C
Color Orange to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
UN Number 3284
Solubility Information Solubility in water: insoluble
Formula Weight 409.27
Percent Purity 97%
Physical Form Crystals or Powder
Chemical Name or Material Diphenyl ditelluride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge
EINECSNumber 250-982-1

RUO – Research Use Only