missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Diselenide 97.0+%, TCI America™

Catalog No. p-7127157
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21865G 5 g
D218625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21865G Supplier TCI America Supplier No. D21865G

Chemical Identifiers

CAS 1666-13-3
Molecular Formula C12H10Se2
Molecular Weight (g/mol) 312.15
MDL Number MFCD00003001
InChI Key YWWZCHLUQSHMCL-UHFFFAOYSA-N
Synonym diphenyl diselenide, diselenide, diphenyl, phenyl diselenide, 1,2-diphenyldiselane, diphenyldiselenide, bis phenylselenide, phenyldiselanyl benzene, diphenyldiselane, ccris 9227, acmc-209duq
PubChem CID 15460
IUPAC Name (phenyldiselanyl)benzene
SMILES [Se]([Se]C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 61°C
Color Yellow
Boiling Point 202°C
Quantity 5 g
UN Number 3283
Formula Weight 312.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyl Diselenide
RTECSNumber JM9152500
TSCA Yes