missing translation for 'onlineSavingsMsg'
Learn More
Learn More
α,α-Diphenyl-4-piperidinomethanol, 98+%, Thermo Scientific™
Specifications
Specifications
| CAS Min % | 98.0 |
| CAS Max % | 100.0 |
| Loss on Drying | 1% max. |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% min. (HPLC) |
| Packaging | Glass bottle |
| Quantity | 5g |
| Merck Index | 13, 895 |
| Formula Weight | 267.37 |
| Percent Purity | 98+% |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes serious eye irritation. Harmful if swallowed. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. |
| EINECSNumber | 204-092-5 |
| RTECSNumber | TN0470000 |
| TSCA | TSCA |
RUO – Research Use Only