missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide

Catalog No. 41595250G
Encompass
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41595250G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41595250G Supplier Sigma Aldrich Supplier No. 41595250G

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide

Specifications

CAS 75980-60-8
Melting Point 88°C to 92°C
Molecular Formula C22H21O2P
Linear Formula (CH3)3C6H2COP(O)(C6H5)2
Quantity 50 g
MDL Number MFCD00192110
Synonym (2,4,6-Trimethylbenzoyl)diphenylphosphine oxide; (Diphenylphosphoryl)(mesityl)methanone; 2,4,6-Trimethylbenzoylphenyl phosphinate
Molecular Weight (g/mol) 348.37
Percent Purity 97%