Learn More
Abcam Inc Diphenhydramine hydrochloride, H1 antagonist, 500MG
Catalog No. 502707326
Encompass
MW 291.82 g/mol, Purity >99%. Potent H receptor antagonist (Ki = 1 nM). Shows antihistaminergic, antiemetic, sedative and hypnotic effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502707326 Supplier Abcam Inc Supplier No. AB120733500MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 291.82 g/mol, Purity >99%. Potent H receptor antagonist (Ki = 1 nM). Shows antihistaminergic, antiemetic, sedative and hypnotic effects.
The product is subject to the following: Abcam Restricted Use Statement