missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipentaerythritol 80.0+%, TCI America™
$33.56 - $78.29
Chemical Identifiers
| CAS | 126-58-9 |
|---|---|
| Molecular Formula | C10H22O7 |
| Molecular Weight (g/mol) | 254.279 |
| MDL Number | MFCD00004691 |
| InChI Key | TXBCBTDQIULDIA-UHFFFAOYSA-N |
| Synonym | dipentaerythritol, dipentek, 2,2'-oxybis methylene bis 2-hydroxymethyl propane-1,3-diol, bis pentaerythritol, 1,3-propanediol, 2,2'-oxybis methylene bis 2-hydroxymethyl, 2,2,2',2'-tetrakis hydroxymethyl-3,3'-oxydipropan-1-ol, 1,3-propanediol, 2,2'-oxybis-methylene, 2-3-hydroxy-2,2-bis hydroxymethyl propoxy methyl-2-hydroxymethyl propane-1,3-diol |
| PubChem CID | 31352 |
| IUPAC Name | 2-[[3-hydroxy-2,2-bis(hydroxymethyl)propoxy]methyl]-2-(hydroxymethyl)propane-1,3-diol |
| SMILES | C(C(CO)(CO)COCC(CO)(CO)CO)O |
Chemical Identifiers
| 126-58-9 | |
| 254.279 | |
| TXBCBTDQIULDIA-UHFFFAOYSA-N | |
| 31352 | |
| C(C(CO)(CO)COCC(CO)(CO)CO)O |
| C10H22O7 | |
| MFCD00004691 | |
| dipentaerythritol, dipentek, 2,2'-oxybis methylene bis 2-hydroxymethyl propane-1,3-diol, bis pentaerythritol, 1,3-propanediol, 2,2'-oxybis methylene bis 2-hydroxymethyl, 2,2,2',2'-tetrakis hydroxymethyl-3,3'-oxydipropan-1-ol, 1,3-propanediol, 2,2'-oxybis-methylene, 2-3-hydroxy-2,2-bis hydroxymethyl propoxy methyl-2-hydroxymethyl propane-1,3-diol | |
| 2-[[3-hydroxy-2,2-bis(hydroxymethyl)propoxy]methyl]-2-(hydroxymethyl)propane-1,3-diol |
Specifications
| 126-58-9 | |
| White | |
| MFCD00004691 | |
| dipentaerythritol, dipentek, 2,2'-oxybis methylene bis 2-hydroxymethyl propane-1,3-diol, bis pentaerythritol, 1,3-propanediol, 2,2'-oxybis methylene bis 2-hydroxymethyl, 2,2,2',2'-tetrakis hydroxymethyl-3,3'-oxydipropan-1-ol, 1,3-propanediol, 2,2'-oxybis-methylene, 2-3-hydroxy-2,2-bis hydroxymethyl propoxy methyl-2-hydroxymethyl propane-1,3-diol | |
| C(C(CO)(CO)COCC(CO)(CO)CO)O | |
| 254.279 | |
| 254.28 | |
| Crystalline Powder |
| 218°C | |
| C10H22O7 | |
| 25 g | |
| TXBCBTDQIULDIA-UHFFFAOYSA-N | |
| 2-[[3-hydroxy-2,2-bis(hydroxymethyl)propoxy]methyl]-2-(hydroxymethyl)propane-1,3-diol | |
| 31352 | |
| ≥80.0% (GC) | |
| Dipentaerythritol |
Safety and Handling
EINECSNumber : (2)-0419
TSCA : Yes