missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipentaerythritol 80.0+%, TCI America™
Specifications
Specifications
| Melting Point | 218°C |
| Color | White |
| Quantity | 25 g |
| Formula Weight | 254.28 |
| Percent Purity | ≥80.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Dipentaerythritol |
Safety and Handling
| EINECSNumber | (2)-0419 |
| TSCA | Yes |