missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ DIPak™ Cartridge

Catalog No. CPDI0NJM1
Change view
Click to view available options
Type:
Cartridge
DI-PAK Purification Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
CPDI0NJM1 Cartridge
CPDI000L1 DI-PAK Purification Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CPDI0NJM1 Supplier MilliporeSigma™ Supplier No. CPDI0NJM1

Produces deionized water directly from tap water

  • “Ion-exchange” and “depth filtration” technology
  • Produces 3–5 L of pure water per day
  • Feed water nature: Tap water
  • Can be easily snapped in and out in seconds without risk of leakage

Specifications

Feedwater Source Tap
Purification Method Ion Exchange, Depth Filtration
For Use With (Equipment) Milli-DI system
Type Cartridge