missing translation for 'onlineSavingsMsg'
Learn More

Diosmin

Catalog No. AAJ6207314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6207314 25 g
AAJ6207306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6207314 Supplier Thermo Scientific Chemicals Supplier No. J6207314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 520-27-4 | C28H32O15 | 608.549 g/mol

Diosmin exhibits anti-proliferative and anti-inflammatory. It is used in the treatment of venous disease, i.e., chronic venous insufficiency (CVI) and hemorrhoidal disease (HD).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in DMSO (50 mg/ml), water (122 mg/ml at 25°C), and ethanol (<1mg/mL at 25°C).

Notes
Protect from heat. Incompatible with heat and oxidizing agents. Store at 4°C.

Chemical Identifiers

CAS 520-27-4
Molecular Formula C28H32O15
Molecular Weight (g/mol) 608.549
MDL Number MFCD00009772
InChI Key GZSOSUNBTXMUFQ-YFAPSIMESA-N
Synonym diosmin, barosmin, diosmine, venosmine, diosmil, flebosten, daflon, diosimin, diosminum, tovene
PubChem CID 5281613
ChEBI CHEBI:4631
IUPAC Name 5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one
SMILES CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=CC(=C4C(=C3)OC(=CC4=O)C5=CC(=C(C=C5)OC)O)O)O)O)O)O)O)O

Specifications

Melting Point 274°C
Color Brown to Yellow
Quantity 25 g
Merck Index 14,3297
Solubility Information Soluble in DMSO (50mg/ml),water (122mg/ml at 25°C),and ethanol (<1mg/ml at 25°C).
Formula Weight 608.5
Physical Form Powder
Chemical Name or Material Diosmin
EINECSNumber 208-289-7
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only