missing translation for 'onlineSavingsMsg'
Learn More

Diosmin 85.0+%, TCI America™

Catalog No. p-7127989
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39085G 5 g
D390825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39085G Supplier TCI America Supplier No. D39085G
Only null left

Chemical Identifiers

CAS 520-27-4
Molecular Formula C28H32O15
Molecular Weight (g/mol) 608.549
MDL Number MFCD00009772
InChI Key GZSOSUNBTXMUFQ-YFAPSIMESA-N
Synonym diosmin, barosmin, diosmine, venosmine, diosmil, flebosten, daflon, diosimin, diosminum, tovene
PubChem CID 5281613
ChEBI CHEBI:4631
IUPAC Name 5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one
SMILES CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=CC(=C4C(=C3)OC(=CC4=O)C5=CC(=C(C=C5)OC)O)O)O)O)O)O)O)O

Specifications

Melting Point 280°C
Quantity 5 g
Formula Weight 608.55
Percent Purity ≥85.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Diosmin