missing translation for 'onlineSavingsMsg'
Learn More

Diosgenin 95.0+%, TCI America™

Catalog No. p-7126742
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D14741G 1 g
D147410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D14741G Supplier TCI America Supplier No. D14741G

Chemical Identifiers

CAS 512-04-9
Molecular Formula C27H42O3
Molecular Weight (g/mol) 414.63
MDL Number MFCD00016887,MFCD01002142
InChI Key WQLVFSAGQJTQCK-VKROHFNGSA-N
Synonym Nitogenin
PubChem CID 46783391
IUPAC Name (1'S,2R,2'S,4'S,5R,7'S,8'R,9'S,12'S,13'R,16'S)-5,7',9',13'-tetramethyl-5'-oxaspiro[oxane-2,6'-pentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosan]-18'-en-16'-ol
SMILES C[C@H]1[C@H]2[C@H](C[C@H]3[C@@H]4CC=C5C[C@@H](O)CC[C@]5(C)[C@H]4CC[C@]23C)O[C@]11CC[C@@H](C)CO1

Specifications

Melting Point 205°C
Color White-Yellow
Quantity 1 g
Formula Weight 414.63
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diosgenin
RTECSNumber WH1450000
TSCA No