missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Diol-Functionalized Silica Gel, Spherical, 40-70 μm

Catalog No. 111012879
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012879 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012879 Supplier MilliporeSigma™ Supelco™ Supplier No. 93981100G

Diol-functionalized silica gel may be used as stationary phase for liquid chromatographic analysis.

Diol-functionalized silica gel is less polar and has lower retention time when compared to normal phase silica gel.

Specifications

Particle Size 40 to 70 μm
Pore Size 110 Å
Grade Standard
Quantity 100 g
Surface Area (Metric) ˜300 m2/g
Description Carbon content, 4.0 to 7.0%
Physical Form Powder
CAS 126850-04-2