missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Dioctyl sulfosuccinate sodium salt
Description
Dioctyl sulfosuccinate sodium salt
Spécifications
Spécifications
| CAS | 577-11-7 |
| Melting Point | 173°C to 179°C (lit.) |
| Molecular Formula | C20H37NaO7S |
| Linear Formula | CH3(CH2)3 CH(C2H5)CH2O2CCH2CH(SO3Na)CO2CH2CH(C2H5)(CH2)3 CH3 |
| Quantity | 5 kg |
| MDL Number | MFCD00012455 |
| Synonym | AOT; Bis(2-ethylhexyl) sulfosuccinate sodium salt; DOSS; Docusate sodium |
| Molecular Weight (g/mol) | 444.56 |
| Percent Purity | ≥97% |