Learn More
Dunwell Tech Inc USB 3.0 Digital Microscope AM73515MT8A- 5MP, 700x - 900x Optical Magnification, Measurement, AMR, Axial Lighting
Catalog No. NC1986114
Encompass
The AM73515MT8A is the USB 3.0 version of the 5MP Dino-Lite Edge Series microscope that has a magnification range of 700x- 900x with built-in axial lighting.Measurements can be performed in different units (inch/mil/mm/um) and types including straight lines, circles, angles, arches grids and more. AMR displays the current magnification on screen with the included software for Windows or Mac OS. Through the use of Flexible LED Control (FLC) using the included software, this model is capable of switching between (or mixing) brightfield and darkfield illumination, revealing details that are very difficult to see under normal light.
Catalog No. NC1986114 Supplier Dunwell Tech Inc Supplier No. AM73515MT8A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The AM73515MT8A is the USB 3.0 version of the 5MP Dino-Lite Edge Series microscope that has a magnification range of 700x- 900x with built-in axial lighting.Measurements can be performed in different units (inch/mil/mm/um) and types including straight lines, circles, angles, arches grids and more. AMR displays the current magnification on screen with the included software for Windows or Mac OS. Through the use of Flexible LED Control (FLC) using the included software, this model is capable of switching between (or mixing) brightfield and darkfield illumination, revealing details that are very difficult to see under normal light.