missing translation for 'onlineSavingsMsg'
Learn More

Dimidium Bromide 98.0+%, TCI America™

Catalog No. p-7126946
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18151G 1 g
D18155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D18151G Supplier TCI America Supplier No. D18151G
Only null left

Chemical Identifiers

CAS 518-67-2
Molecular Formula C20H18BrN3
Molecular Weight (g/mol) 380.29
MDL Number MFCD00011757
InChI Key MQOKYEROIFEEBH-UHFFFAOYSA-N
Synonym dimidium bromide, trypadine, 3,8-diamino-5-methyl-6-phenylphenanthridinium bromide, phenanthridinium 1553, phenanthridinium compound 1553, phenanthridinium, 3,8-diamino-5-methyl-6-phenyl-, bromide, 2,7-diamino-10-methyl-9-phenylphenanthridinium bromide, 3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide, phenanthridium 1553
PubChem CID 68207
IUPAC Name 3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide
SMILES [Br-].C[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12

Specifications

Color Purple-Red
Quantity 1 g
Formula Weight 380.29
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Dimidium Bromide
RTECSNumber SF7960500
TSCA No