missing translation for 'onlineSavingsMsg'
Learn More

Dimidium bromide, 98%

CAS: 518-67-2 | C20H18BrN3 | 380.29 g/mol

Supplier:  Thermo Scientific Chemicals 215342500

 View more versions of this product

Catalog No. AC215342500


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dimidium bromide
518-67-2
Purple to Red
97.5% min. (on dry substance) (Argentometry)
C20H18BrN3
250 mg
09, 3257
Solubility in water: soluble
[Br-].C[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
380.29
380.28
Powder
98%
230°C
Authentic
Glass bottle
MFCD00011757
22, IV, 5515
dimidium bromide, trypadine, 3,8-diamino-5-methyl-6-phenylphenanthridinium bromide, phenanthridinium 1553, phenanthridinium compound 1553, phenanthridinium, 3,8-diamino-5-methyl-6-phenyl-, bromide, 2,7-diamino-10-methyl-9-phenylphenanthridinium bromide, 3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide, phenanthridium 1553
MQOKYEROIFEEBH-UHFFFAOYSA-N
5-methyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide
68207
98%

Chemical Identifiers

518-67-2
380.29
MQOKYEROIFEEBH-UHFFFAOYSA-N
68207
[Br-].C[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
C20H18BrN3
MFCD00011757
dimidium bromide, trypadine, 3,8-diamino-5-methyl-6-phenylphenanthridinium bromide, phenanthridinium 1553, phenanthridinium compound 1553, phenanthridinium, 3,8-diamino-5-methyl-6-phenyl-, bromide, 2,7-diamino-10-methyl-9-phenylphenanthridinium bromide, 3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide, phenanthridium 1553
5-methyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 208-256-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only