missing translation for 'onlineSavingsMsg'
Learn More

Dimidium Bromide, 97%, Spectrum™ Chemical

Catalog No. p-7165493
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
100 g
1 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610253 100 mg Amber Glass Bottle
18610255 500 mg Amber Glass Bottle
18610250 1 g Amber Glass Bottle
18610252 100 g Amber Glass Bottle
18610251 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18610253 Supplier Spectrum Chemical Mfg Cor Supplier No. D2499100MG
Only null left

Chemical Identifiers

CAS 518-67-2

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 100 mg
Percent Purity 0.97
Grade Reagent

CAUTION: For use by qualified personnel for research and development only. Read and understand the label and Safety Data Sheet (SDS) prior to use.