missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyltitanocene (5% in Tetrahydrofuran/Toluene), TCI America™
Supplier: TCI America D4100100G
Specifications
| Dimethyltitanocene (5% in Tetrahydrofuran/Toluene) | |
| C12H20Ti | |
| 100 g | |
| bis cyclopentadienyl dimethyltitanium, dimethyltitanocene in tetrahydrofuran/toluene | |
| C.C.C1C=CC=C1.C1C=CC=C1.[Ti] | |
| 212.159 | |
| 208.13 |
| 1271-66-5 | |
| MFCD02093882 | |
| 1993 | |
| CRKLPNOLYBORCP-UHFFFAOYSA-N | |
| cyclopenta-1,3-diene;methane;titanium | |
| 131674729 | |
| Liquid |
Chemical Identifiers
| 1271-66-5 | |
| 212.159 | |
| CRKLPNOLYBORCP-UHFFFAOYSA-N | |
| 131674729 | |
| C.C.C1C=CC=C1.C1C=CC=C1.[Ti] |
| C12H20Ti | |
| MFCD02093882 | |
| bis cyclopentadienyl dimethyltitanium, dimethyltitanocene in tetrahydrofuran/toluene | |
| cyclopenta-1,3-diene;methane;titanium |
Safety and Handling
TSCA : No
Recommended Storage : Freezer