missing translation for 'onlineSavingsMsg'
Learn More

Dimethyltin oxide

Catalog No. p-7080329
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA7116404 2 g
AA7116409 10 g
AA7116418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA7116404 Supplier Thermo Scientific Chemicals Supplier No. 07116404
Only null left

CAS: 2273-45-2 | C2H6OSn | 164.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2273-45-2
Molecular Formula C2H6OSn
Molecular Weight (g/mol) 164.78
MDL Number MFCD00013838
InChI Key WNVQCJNZEDLILP-UHFFFAOYSA-N
Synonym dimethyltin oxide, stannane, dimethyloxo, dimethyloxostannane, tin, dimethyloxo, dimethyltinoxide, dimethyl oxo tin, dimethyl tin oxide, unii-409qf23wo7, dimethyl oxo stannane, oxodimethyltin
PubChem CID 75277
IUPAC Name dimethyl(oxo)tin
SMILES C[Sn](C)=O

Specifications

Melting Point (decomposition)
Quantity 2 g
UN Number UN3146
Formula Weight 166.77
Physical Form Powder
Chemical Name or Material Dimethyltin oxide
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
EINECSNumber 218-881-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only