missing translation for 'onlineSavingsMsg'
Learn More

Dimethyltin Dichloride 99.0+%, TCI America™

Catalog No. p-7126686
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D133825G 25 g
D1338100G 100 g
D1338500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D133825G Supplier TCI America Supplier No. D133825G
Only null left

Chemical Identifiers

CAS 753-73-1
Molecular Formula C2H6Cl2Sn
Molecular Weight (g/mol) 219.68
MDL Number MFCD00000517
InChI Key PKKGKUDPKRTKLJ-UHFFFAOYSA-L
Synonym dimethyltin dichloride, dimethyldichlorotin, stannane, dichlorodimethyl, dichlorodimethyltin, dimethyltindichloride, dimethyldichlorostannane, dichloro dimethyl stannane, tin, dimethyl-, dichloride, cotin 210, dichlorid dimethylcinicity
PubChem CID 12955
IUPAC Name dichloro(dimethyl)stannane
SMILES C[Sn](C)(Cl)Cl

Specifications

Melting Point 107°C
Color White-Yellow
Boiling Point 180°C
Quantity 25 g
UN Number 2928
Formula Weight 219.68
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyltin Dichloride
EINECSNumber (2)-2247
RTECSNumber WH7245000
TSCA Yes