missing translation for 'onlineSavingsMsg'
Learn More

Dimethyltin dichloride, 95%, Thermo Scientific™

$109.15 - $1,252.54

Chemical Identifiers

CAS 753-73-1
Molecular Formula C2H6Cl2Sn
Molecular Weight (g/mol) 219.66
MDL Number MFCD00000517
InChI Key PKKGKUDPKRTKLJ-UHFFFAOYSA-L
Synonym dimethyltin dichloride, dimethyldichlorotin, stannane, dichlorodimethyl, dichlorodimethyltin, dimethyltindichloride, dimethyldichlorostannane, dichloro dimethyl stannane, tin, dimethyl-, dichloride, cotin 210, dichlorid dimethylcinicity
PubChem CID 12955
IUPAC Name dichloro(dimethyl)stannane
SMILES C[Sn](C)(Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC347820050
View Documents Promotion Details
N/A 5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC347821000
View Documents Promotion Details
N/A 100g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

753-73-1
219.66
PKKGKUDPKRTKLJ-UHFFFAOYSA-L
12955
C[Sn](C)(Cl)Cl
C2H6Cl2Sn
MFCD00000517
dimethyltin dichloride, dimethyldichlorotin, stannane, dichlorodimethyl, dichlorodimethyltin, dimethyltindichloride, dimethyldichlorostannane, dichloro dimethyl stannane, tin, dimethyl-, dichloride, cotin 210, dichlorid dimethylcinicity
dichloro(dimethyl)stannane

Specifications

753-73-1
Authentic
Glass bottle
(CH3)2Sn(Cl)2
dimethyltin dichloride, dimethyldichlorotin, stannane, dichlorodimethyl, dichlorodimethyltin, dimethyltindichloride, dimethyldichlorostannane, dichloro dimethyl stannane, tin, dimethyl-, dichloride, cotin 210, dichlorid dimethylcinicity
PKKGKUDPKRTKLJ-UHFFFAOYSA-L
dichloro(dimethyl)stannane
12955
95%
188.0°C to 190.0°C
94% min. (Argentometry)
C2H6Cl2Sn
MFCD00000517
Solubility in water: 20g/L (20°C). Other solubilities: 0.1 g/mL in methanol
C[Sn](C)(Cl)Cl
219.66
219.66
Dimethyltin dichloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Suspected of damaging the unborn child.
Causes damage to organs through prolonged or repeated

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Danger

EINECSNumber : 212-039-2

SDS
Documents

Documents

RUO – Research Use Only