missing translation for 'onlineSavingsMsg'
Learn More

Dimethylsulfamoyl Chloride, 99%

CAS: 13360-57-1 | C2H6ClNO2S | 143.59 g/mol

$142.94 - $142.94

Chemical Identifiers

CAS 13360-57-1
Molecular Formula C2H6ClNO2S
Molecular Weight (g/mol) 143.59
MDL Number MFCD00007425
InChI Key JFCHSQDLLFJHOA-UHFFFAOYSA-N
Synonym dimethylsulfamoyl chloride, sulfamoyl chloride, dimethyl, dimethylaminosulfonyl chloride, dimethylsulfamoylchloride, dimethylsulphamoyl chloride, dimethylamidosulfonyl chloride, n,n-dimethylsulfamyl chloride, dimethylsulfamyl chloride, sulfamoyl chloride, n,n-dimethyl, dimethyl sulfamoyl chloride
PubChem CID 83372
IUPAC Name N,N-dimethylsulfamoyl chloride
SMILES CN(C)S(Cl)(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC408590250
SDS View Documents
Thermo Scientific Chemicals
408590250
25 g Glass bottle
Each for $142.94
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13360-57-1
143.59
JFCHSQDLLFJHOA-UHFFFAOYSA-N
83372
CN(C)S(Cl)(=O)=O
C2H6ClNO2S
MFCD00007425
dimethylsulfamoyl chloride, sulfamoyl chloride, dimethyl, dimethylaminosulfonyl chloride, dimethylsulfamoylchloride, dimethylsulphamoyl chloride, dimethylamidosulfonyl chloride, n,n-dimethylsulfamyl chloride, dimethylsulfamyl chloride, sulfamoyl chloride, n,n-dimethyl, dimethyl sulfamoyl chloride
N,N-dimethylsulfamoyl chloride

Specifications

13360-57-1
1.3370g/mL
94°C
Glass bottle
1.4510 to 1.453
MFCD00007425
04, 84
1.337
Solubility in water: decomposes in water
CN(C)S(Cl)(=O)=O
143.59
143.59
Dimethylsulfamoyl chloride
-13°C
114°C (75.0 mmHg)
98.5% min. (GC)
C2H6ClNO2S
(CH3)2NSO2Cl
25 g
09,187
dimethylsulfamoyl chloride, sulfamoyl chloride, dimethyl, dimethylaminosulfonyl chloride, dimethylsulfamoylchloride, dimethylsulphamoyl chloride, dimethylamidosulfonyl chloride, n,n-dimethylsulfamyl chloride, dimethylsulfamyl chloride, sulfamoyl chloride, n,n-dimethyl, dimethyl sulfamoyl chloride
JFCHSQDLLFJHOA-UHFFFAOYSA-N
N,N-dimethylsulfamoyl chloride
83372
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
May cause cancer.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 236-412-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only