missing translation for 'onlineSavingsMsg'
Learn More

Dimethylphenylsilanol sodium salt, 97%

Catalog No. p-7030407
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2845703 1 g
AAH2845706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2845703 Supplier Thermo Scientific Chemicals Supplier No. H2845703
Only null left

CAS: 7646-75-5 | C8H11NaOSi | 174.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7646-75-5
Molecular Formula C8H11NaOSi
Molecular Weight (g/mol) 174.25
MDL Number MFCD09266151
InChI Key NPZJKOMUCXNLBN-UHFFFAOYSA-N
Synonym dimethylphenylsilanol sodium salt, sodium dimethyl phenyl silanolate, sodium dimethylphenylsilanolate, amtsi046, sodium phenyldimethylsilanolate, dimethyl phenyl sodiooxy silane, sodium phenyl dimethylsilanolate
PubChem CID 23713317
IUPAC Name sodium;dimethyl-oxido-phenylsilane
SMILES C[Si](C)(C1=CC=CC=C1)[O-].[Na+]

Specifications

Quantity 1 g
Formula Weight 174.25
Percent Purity 97%
Chemical Name or Material Dimethylphenylsilanol sodium salt
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only