missing translation for 'onlineSavingsMsg'
Learn More

Dimethylphenylsilane 97.0+%, TCI America™

Catalog No. p-7127164
Change view
Click to view available options
Quantity:
25 mL
5 mL
Catalog No. Quantity
D21965ML 5 mL
D219625ML 25 mL
2 options

Catalog No. D21965ML

Supplier: TCI America D21965ML

Only null left

Chemical Identifiers

CAS 766-77-8
Molecular Formula C8H11Si
Molecular Weight (g/mol) 135.261
MDL Number MFCD00008257
InChI Key OIKHZBFJHONJJB-UHFFFAOYSA-N
Synonym dimethylphenylsilane, dimethyl phenyl silane, phenyldimethylsilane, silane, dimethylphenyl, benzene, dimethylsilyl, dimethylphenyl silane, c8h12si, dimethyl phenyl silyl, dimethyl-phenylsilicon
PubChem CID 6327656
IUPAC Name dimethyl(phenyl)silicon
SMILES C[Si](C)C1=CC=CC=C1

Specifications

Boiling Point 158°C
Color Yellow
Physical Form Liquid
Quantity 5 mL
UN Number 1993
Formula Weight 136.27
Percent Purity ≥97.0% (GC)
Chemical Name or Material Dimethylphenylsilane