missing translation for 'onlineSavingsMsg'
Learn More

Dimethylphenylsilane, 97%

Catalog No. AAL0455818
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0455818 50 g
AAL0455809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0455818 Supplier Thermo Scientific Chemicals Supplier No. L0455818
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 766-77-8 | C8H11Si | 135.261 g/mol

Dimethylphenylsilane is a reagent for enol ether synthesis. It acts as a catalyst. Also used as a precursor in Optical Emission Spectroscopy of Plasma Deposition Processes. It can react with vinylbenzene to produce (b-Phenyl-ethyl)-dimethylphenyl-silan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylphenylsilane is a reagent for enol ether synthesis. It acts as a catalyst. Also used as a precursor in Optical Emission Spectroscopy of Plasma Deposition Processes. It can react with vinylbenzene to produce (b-Phenyl-ethyl)-dimethylphenyl-silan.

Solubility
Not miscible in water.

Notes
Moisture sensitive. Incompatible materials are Strong oxidizing agents, Strong acids.

Chemical Identifiers

CAS 766-77-8
Molecular Formula C8H11Si
Molecular Weight (g/mol) 135.261
MDL Number MFCD00008257
InChI Key OIKHZBFJHONJJB-UHFFFAOYSA-N
Synonym dimethylphenylsilane, dimethyl phenyl silane, phenyldimethylsilane, silane, dimethylphenyl, benzene, dimethylsilyl, dimethylphenyl silane, c8h12si, dimethyl phenyl silyl, dimethyl-phenylsilicon
PubChem CID 6327656
IUPAC Name dimethyl(phenyl)silicon
SMILES C[Si](C)C1=CC=CC=C1

Specifications

Boiling Point 156°C to 157°C
Melting Point -124°C
Quantity 50 g
Flash Point 35°C (95°F)
Refractive Index 1.497
Density 0.879
UN Number UN1993
Beilstein 2204906
Solubility Information Not miscible in water.
Formula Weight 136.27
Percent Purity 97%
Chemical Name or Material Dimethylphenylsilane
Show More Show Less
Hazard Category H226-H319-H335
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 212-170-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only