missing translation for 'onlineSavingsMsg'
Learn More

Dimethylphenacylsulfonium Tetrafluoroborate 98.0+%, TCI America™

$149.32 - $459.68

Chemical Identifiers

CAS 24806-57-3
Molecular Formula C10H13BF4OS
Molecular Weight (g/mol) 268.077
MDL Number MFCD00191352
InChI Key NYDZUKNJLPHOOF-UHFFFAOYSA-N
PubChem CID 2737123
IUPAC Name dimethyl(phenacyl)sulfanium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.C[S+](C)CC(=O)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D22311G
View Documents
TCI America
D22311G
1 g N/A
Only null left
 
D22315G
View Documents
TCI America
D22315G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

24806-57-3
268.077
NYDZUKNJLPHOOF-UHFFFAOYSA-N
dimethyl(phenacyl)sulfanium;tetrafluoroborate
C10H13BF4OS
MFCD00191352
2737123
[B-](F)(F)(F)F.C[S+](C)CC(=O)C1=CC=CC=C1

Specifications

24806-57-3
White
MFCD00191352
1759
[B-](F)(F)(F)F.C[S+](C)CC(=O)C1=CC=CC=C1
268.077
268.08
Crystalline Powder
170°C
C10H13BF4OS
1 g
NYDZUKNJLPHOOF-UHFFFAOYSA-N
dimethyl(phenacyl)sulfanium;tetrafluoroborate
2737123
≥98.0% (T)
Dimethylphenacylsulfonium Tetrafluoroborate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'