missing translation for 'onlineSavingsMsg'
Learn More

Dimethyloctadecylchlorosilane 97.0+%, TCI America™

Catalog No. D156025ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D156025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D156025ML Supplier TCI America Supplier No. D156025ML
Only null left

Chemical Identifiers

CAS 18643-08-8
Molecular Formula C20H43ClSi
Molecular Weight (g/mol) 347.10
MDL Number MFCD00000504
InChI Key GZGREZWGCWVAEE-UHFFFAOYSA-N
Synonym chlorodimethyloctadecylsilane, dimethyloctadecylchlorosilane, chlorodimethyl octadecyl silane, octadecyldimethylchlorosilane, silane, chlorodimethyloctadecyl, n-octadecyldimethylchlorosilane, chloro dimethyl octadecylsilane, chlorodimethyl-n-octadecylsilane, dimethyl-n-octadecylchlorosilane, stearyldimethylchlorosilane
PubChem CID 87738
IUPAC Name chlorodimethyloctadecylsilane
SMILES CCCCCCCCCCCCCCCCCC[Si](C)(C)Cl

Specifications

Color White-Yellow
Quantity 25 mL
UN Number 2987
Formula Weight 347.10
Percent Purity ≥97.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material Dimethyloctadecylchlorosilane
EINECSNumber (2)-2041
TSCA Yes