missing translation for 'onlineSavingsMsg'
Learn More

Dimethylglyoxime 98.0+%, TCI America™

Catalog No. D0729500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0729500G 500 g
D072925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0729500G Supplier TCI America Supplier No. D0729500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 95-45-4
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
MDL Number MFCD00002117
InChI Key JGUQDUKBUKFFRO-GGWOSOGESA-N
Synonym biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky
PubChem CID 5356010
IUPAC Name (E)-N-[(3E)-3-(hydroxyimino)butan-2-ylidene]hydroxylamine
SMILES C\C(=N/O)\C(\C)=N\O

Specifications

Melting Point 240°C
Color White
Quantity 500 g
Formula Weight 116.12
Percent Purity ≥98.0% (W)
Physical Form Crystalline Powder
Chemical Name or Material Dimethylglyoxime
EINECSNumber (2)-0541
RTECSNumber EK2975000
TSCA Yes