missing translation for 'onlineSavingsMsg'
Learn More

Dimethylglyoxime, disodium salt octahydrate, 99%

Catalog No. AC408340010
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC408340010 1 kg Glass bottle
AC408341000 100 g Glass bottle
AC408342500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC408340010 Supplier Thermo Scientific Chemicals Supplier No. 408340010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dimethylglyoxime, disodium salt octahydrate, 99%, C4H6N2Na2O2.8H2O, CAS Number-75006-64-3, c4h6n2o2.2na.8h2o, dimethylglyoxime di-sodium salt octahydrate, dimethylglyoxime disodium salt octahydrate, sodium dimethylglyoxime octahydrate, 1kg, White to Beige, 262-523-2, 304.19 | CAS: 75006-64-3 | C4H22N2Na2O10 | 304.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75006-64-3
Molecular Formula C4H22N2Na2O10
Molecular Weight (g/mol) 304.20
MDL Number MFCD00150288
InChI Key IJJDRGUPRATWQX-UHFFFAOYSA-L
Synonym dimethylglyoxime disodium salt octahydrate, c4h6n2o2.2na.8h2o, sodium dimethylglyoxime octahydrate, dimethylglyoxime di-sodium salt octahydrate
PubChem CID 71310106
SMILES O.O.O.O.O.O.O.O.[Na+].[Na+].CC(=N[O-])C(C)=N[O-]

Specifications

Melting Point >300.0°C
Color White to Beige
Loss on Drying 46% to 49%
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3C(NONa)C(NONa)CH3·8H2O
Quantity 1 kg
Solubility Information (50mg/ml,50% ethanol) Clear solution
IUPAC Name disodium oxido[3-(oxidoimino)butan-2-ylidene]amine octahydrate
Formula Weight 304.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Dimethylglyoxime, disodium salt octahydrate, 99%
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 262-523-2
TSCA TSCA

RUO – Research Use Only