missing translation for 'onlineSavingsMsg'
Learn More

Dimethylglyoxime, ACS Grade, LabChem™

Catalog No. LC135969
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC135969 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC135969 Supplier LabChem Supplier No. LC135969

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 95-45-4
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
MDL Number MFCD00002117
InChI Key JGUQDUKBUKFFRO-GGWOSOGESA-N
Synonym biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky
PubChem CID 5356010
IUPAC Name (E)-N-[(3E)-3-(hydroxyimino)butan-2-ylidene]hydroxylamine
SMILES C\C(=N/O)\C(\C)=N\O

Specifications

CAS Max % 100
Melting Point 240°C
Color White
Packaging Poly Bottle
Linear Formula CH3C(=NOH)C(=NOH)CH3
Quantity 100 g
Solubility Information Soluble in water
Formula Weight 116.12
Grade ACS
Physical Form Powder
Chemical Name or Material Dimethylglyoxime
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
May form combustible dust concentrations in air.
EINECSNumber 202-420-1
Recommended Storage Room Temperature