missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylglyoxime, 99+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 239.0°C to 242.0°C |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99+% |
| Packaging | Plastic bottle |
| Linear Formula | CH3C(=NOH)C(=NOH)CH3 |
| Quantity | 100 g |
| Beilstein | 01,772 |
| Merck Index | 15,327 |
| Solubility Information | Solubility in water: 0,6g/L 20°C. Other solubilities: soluble in alcohol,ether,pyridine and acetone,slightly soluble in benzene |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Flammable solid. Toxic if swallowed. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF SWALLOWED: Immediate |
| EINECSNumber | 202-420-1 |
| RTECSNumber | EK2975000 |
| TSCA | TSCA |
RUO – Research Use Only