Learn More
Dimethylglyoxime, 99%
CAS: 95-45-4 | C4H8N2O2 | 116.12 g/mol
Supplier: Thermo Scientific Chemicals A128100E
Description
Dimethylglyoxime is a well known reagent in analytical chemistry. It is used for the qualitative and quantitative determination of nickel by gravimetric analysis. It also finds application in the separation of palladium from tin, gold, rhodium and iridium. It is also useful in the detection of bismuth.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDimethylglyoxime is a well known reagent in analytical chemistry. It is used for the qualitative and quantitative determination of nickel by gravimetric analysis. It also finds application in the separation of palladium from tin, gold, rhodium and iridium. It is also useful in the detection of bismuth.
Solubility
Soluble in alcohol, ether and solutions of alkalies. Insoluble in water.
Notes
Incompatible with oxidizing agents.
Specifications
| Dimethylglyoxime | |
| ∼240°C (decomposition) | |
| MFCD00002117 | |
| 506731 | |
| biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky | |
| JGUQDUKBUKFFRO-GGWOSOGESA-N | |
| N-[(E)-3-nitrosobut-2-en-2-yl]hydroxylamine | |
| 5356010 | |
| 99% |
| 95-45-4 | |
| C4H8N2O2 | |
| 2500 g | |
| 14,3246 | |
| Soluble in alcohol,ether and solutions of alkalies. Insoluble in water. | |
| C\C(=N/O)\C(\C)=N\O | |
| 116.12 | |
| 116.12 |
Chemical Identifiers
| 95-45-4 | |
| 116.12 | |
| JGUQDUKBUKFFRO-GGWOSOGESA-N | |
| 5356010 | |
| C\C(=N/O)\C(\C)=N\O |
| C4H8N2O2 | |
| MFCD00002117 | |
| biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky | |
| N-[(E)-3-nitrosobut-2-en-2-yl]hydroxylamine |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P210-P240-P241-P264b-P270-P280-P301+P310-P330-P370+P378q-P501c
H228-H301-H500
DOTInformation : Hazard Class: 4.1; Packaging Group: III
EINECSNumber : 202-420-1
RTECSNumber : EK2975000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only