Learn More
Dimethylglyoxime, 99+%, ACS reagent
CAS: 95-45-4 | C4H8N2O2 | 116.12 g/mol
Supplier: Thermo Scientific Chemicals 408331000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Dimethylglyoxime | |
| 239.0°C to 242.0°C | |
| 0.05% max. | |
| 99+% | |
| C4H8N2O2 | |
| MFCD00002117 | |
| 01, 772 | |
| biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky | |
| Solubility in water: 0,6g/L 20°C. Other solubilities: soluble in alcohol,ether,pyridine and acetone,slightly soluble in benzene | |
| C\C(=N/O)\C(\C)=N\O | |
| 116.12 | |
| 116.12 | |
| ACS Reagent | |
| Crystalline Powder |
| 95-45-4 | |
| White | |
| Authentic | |
| Plastic bottle | |
| CH3C(=NOH)C(=NOH)CH3 | |
| 100 g | |
| 15, 3270 | |
| >70°C | |
| JGUQDUKBUKFFRO-GGWOSOGESA-N | |
| N-[(E)-3-nitrosobut-2-en-2-yl]hydroxylamine | |
| 5356010 | |
| 99+% | |
| 0.02% max. (in ethanol) |
Chemical Identifiers
| 95-45-4 | |
| 116.12 | |
| JGUQDUKBUKFFRO-GGWOSOGESA-N | |
| 5356010 |
| C4H8N2O2 | |
| MFCD00002117 | |
| biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky | |
| C\C(=N/O)\C(\C)=N\O |
Safety and Handling
GHS H Statement
Flammable solid.
Toxic if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediate
GHS Signal Word: Danger
EINECSNumber : 202-420-1
RTECSNumber : EK2975000
TSCA : TSCA
RUO – Research Use Only