missing translation for 'onlineSavingsMsg'
Learn More

Dimethylethylhexadecylammonium bromide, 99%

CAS: 124-03-8 | C20H44BrN | 378.483 g/mol

$166.68 - $487.96

Chemical Identifiers

CAS 124-03-8
Molecular Formula C20H44BrN
Molecular Weight (g/mol) 378.483
MDL Number MFCD00011774
InChI Key VUFOSBDICLTFMS-UHFFFAOYSA-M
Synonym Cetyldimethylethylammonium bromide
PubChem CID 31280
IUPAC Name ethyl-hexadecyl-dimethylazanium;bromide
SMILES CCCCCCCCCCCCCCCC[N+](C)(C)CC.[Br-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC211871000
View Documents Promotion Details
N/A 100 g Glass bottle
Each for $166.68
Only null left
 
AC211875000
View Documents Promotion Details
N/A 500 g Glass bottle
Each for $487.96
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Chemical Identifiers

124-03-8
378.483
VUFOSBDICLTFMS-UHFFFAOYSA-M
31280
CCCCCCCCCCCCCCCC[N+](C)(C)CC.[Br-]
C20H44BrN
MFCD00011774
Cetyldimethylethylammonium bromide
ethyl-hexadecyl-dimethylazanium;bromide

Specifications

White
Powder
Authentic
C20H44BrN
MFCD00011774
15, 2028
VUFOSBDICLTFMS-UHFFFAOYSA-M
ethyl-hexadecyl-dimethylazanium;bromide
31280
≥98.5%
124-03-8
6 to 8 (1% soln.)
99%
CH3(CH2)15N(CH3)2(C2H5)Br
04, III, 425
Cetyldimethylethylammonium bromide
CCCCCCCCCCCCCCCC[N+](C)(C)CC.[Br-]
378.483
378.48
Dimethylethylhexadecylammonium bromide, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 204-672-8

RTECSNumber : BQ5425000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only