missing translation for 'onlineSavingsMsg'
Learn More

Dimethylethoxyvinylsilane, 97%

Catalog No. p-7046888
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1669914 25 g
AAL1669922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1669914 Supplier Thermo Scientific Chemicals Supplier No. L1669914

CAS: 5356-83-2 | C6H14OSi | 130.262 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5356-83-2
Molecular Formula C6H14OSi
Molecular Weight (g/mol) 130.262
MDL Number MFCD00009070
InChI Key JEWCZPTVOYXPGG-UHFFFAOYSA-N
Synonym ethoxydimethylvinylsilane, dimethylethoxyvinylsilane, ethoxydimethyl vinyl silane, silane, ethenylethoxydimethyl, silane, ethoxydimethylvinyl, vinyldimethylethoxysilane, ethenylethoxydimethyl silane, ethoxy dimethyl vinylsilane, dimethylvinylethoxysilane, dimethyl ethenyl silyloxyethane
PubChem CID 79315
IUPAC Name ethenyl-ethoxy-dimethylsilane
SMILES CCO[Si](C)(C)C=C

Specifications

Boiling Point 99°C
Quantity 25 g
UN Number UN1993
Beilstein 1743477
Flash Point −3°C (26°F)
Formula Weight 130.26
Percent Purity 97%
Refractive Index 1.398
Sensitivity Moisture sensitive
Density 0.79
Chemical Name or Material Dimethylethoxyvinylsilane
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 226-341-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only