missing translation for 'onlineSavingsMsg'
Learn More

Dimethylethoxysilane, 94%

Catalog No. AC392001000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC392001000 100 g Glass bottle
AC392000050 5 g Glass bottle
AC392000250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC392001000 Supplier Thermo Scientific Chemicals Supplier No. 392001000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14857-34-2 | C4H12OSi | 104.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

Chemical Identifiers

CAS 14857-34-2
Molecular Formula C4H12OSi
Molecular Weight (g/mol) 104.22
MDL Number MFCD00026750
InChI Key DRUOQOFQRYFQGB-UHFFFAOYSA-N
Synonym ethoxydimethylsilane, dimethylethoxysilane, silane, ethoxydimethyl, dimethyl ethoxysilane, ethoxy dimethyl silicon, dimethylethoxysilane, ammonia-free, ethoxydimethylsilyl, ethoxydimethyl-silan, sgqhhgcihcup@, 4-04-00-03991 beilstein handbook reference
PubChem CID 6328550
IUPAC Name ethoxy(dimethyl)silicon
SMILES CCO[Si](C)C

Specifications

Color Colorless
Boiling Point 54.0°C
Flash Point −5°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Refractive Index 1.3670 to 1.3690 (20°C, 589nm)
Linear Formula CH3CH2OSiH(CH3)2
Quantity 100 g
Solubility Information Solubility in water: reacts
Formula Weight 104.22
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Dimethylethoxysilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 238-921-7

RUO – Research Use Only