missing translation for 'onlineSavingsMsg'
Learn More

Dimethylethoxysilane, 94%

Catalog No. AAL1647614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1647614 25 g
AAL1647622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1647614 Supplier Thermo Scientific Chemicals Supplier No. L1647614

CAS: 14857-34-2 | C4H11OSi | 103.216 g/mol

Dimethylethoxysilane (DMES), a volatile liquid, is used by NASA to waterproof the heatprotective silica tiles and blankets on the space shuttle. DMES readily produces a variety of products in the presence of small amounts of moisture or bases. DMES readily produces a variety of products in the presence of small amounts of moisture or bases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylethoxysilane (DMES), a volatile liquid, is used by NASA to waterproof the heatprotective silica tiles and blankets on the space shuttle. DMES readily produces a variety of products in the presence of small amounts of moisture or bases. DMES readily produces a variety of products in the presence of small amounts of moisture or bases.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water.

Chemical Identifiers

CAS 14857-34-2
Molecular Formula C4H11OSi
Molecular Weight (g/mol) 103.216
MDL Number MFCD00026750
InChI Key DRUOQOFQRYFQGB-UHFFFAOYSA-N
Synonym ethoxydimethylsilane, dimethylethoxysilane, silane, ethoxydimethyl, dimethyl ethoxysilane, ethoxy dimethyl silicon, dimethylethoxysilane, ammonia-free, ethoxydimethylsilyl, ethoxydimethyl-silan, sgqhhgcihcup@, 4-04-00-03991 beilstein handbook reference
PubChem CID 6328550
IUPAC Name ethoxy(dimethyl)silicon
SMILES CCO[Si](C)C

Specifications

Boiling Point 55°C
Quantity 25 g
UN Number UN1993
Beilstein 1731481
Flash Point 15°C (59°F)
Solubility Information Hydrolyzes in water.
Formula Weight 104.23
Percent Purity 94%
Refractive Index 1.368
Sensitivity Moisture sensitive
Density 0.757
Chemical Name or Material Dimethylethoxysilane, ammonia-free
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 238-921-7
RTECSNumber VV4090000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only