missing translation for 'onlineSavingsMsg'
Learn More

Dimethyldioctadecylammonium Bromide 98.0+%, TCI America™

Catalog No. p-7127041
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D197525G 25 g
D1975250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D197525G Supplier TCI America Supplier No. D197525G
Only null left

Chemical Identifiers

CAS 3700-67-2
Molecular Formula C38H80BrN
Molecular Weight (g/mol) 630.969
MDL Number MFCD00041975
InChI Key PSLWZOIUBRXAQW-UHFFFAOYSA-M
Synonym dimethyldioctadecylammonium bromide, n,n-dimethyl-n-octadecyloctadecan-1-aminium bromide, dimethyldistearylammonium bromide, 1-octadecanaminium, n,n-dimethyl-n-octadecyl-, bromide, unii-z5t47r065a, dsab, distearyldimethylammonium bromide, dimethyldioctadecylazanium bromide, distearyl dimethyl ammonium bromide, 1-octadecanaminium, n,n-dimethyl-n-octadecyl-, bromide 1:1
PubChem CID 77293
IUPAC Name dimethyl(dioctadecyl)azanium;bromide
SMILES CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCCCC.[Br-]

Specifications

Melting Point 155°C
Color White
Quantity 25 g
Formula Weight 630.97
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyldioctadecylammonium Bromide
EINECSNumber (2)-0184&(9)-1971
TSCA Yes