Learn More
Dimethyldioctadecylammonium bromide, 99+%
CAS: 3700-67-2 | C38H80BrN | 630.96 g/mol
$148.37 - $148.37
Chemical Identifiers
| CAS | 3700-67-2 |
|---|---|
| Molecular Formula | C38H80BrN |
| Molecular Weight (g/mol) | 630.96 |
| MDL Number | MFCD00041975 |
| InChI Key | PSLWZOIUBRXAQW-UHFFFAOYSA-M |
| Synonym | dimethyldioctadecylammonium bromide, n,n-dimethyl-n-octadecyloctadecan-1-aminium bromide, dimethyldistearylammonium bromide, 1-octadecanaminium, n,n-dimethyl-n-octadecyl-, bromide, unii-z5t47r065a, dsab, distearyldimethylammonium bromide, dimethyldioctadecylazanium bromide, distearyl dimethyl ammonium bromide, 1-octadecanaminium, n,n-dimethyl-n-octadecyl-, bromide 1:1 |
| PubChem CID | 77293 |
| IUPAC Name | dimethyl(dioctadecyl)azanium;bromide |
| SMILES | CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCCCC.[Br-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC408260050
|
Thermo Scientific Chemicals
408260050 |
5 g | Glass bottle |
Each for $148.37
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystChemical Identifiers
| 3700-67-2 | |
| 630.96 | |
| PSLWZOIUBRXAQW-UHFFFAOYSA-M | |
| 77293 | |
| CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCCCC.[Br-] |
| C38H80BrN | |
| MFCD00041975 | |
| dimethyldioctadecylammonium bromide, n,n-dimethyl-n-octadecyloctadecan-1-aminium bromide, dimethyldistearylammonium bromide, 1-octadecanaminium, n,n-dimethyl-n-octadecyl-, bromide, unii-z5t47r065a, dsab, distearyldimethylammonium bromide, dimethyldioctadecylazanium bromide, distearyl dimethyl ammonium bromide, 1-octadecanaminium, n,n-dimethyl-n-octadecyl-, bromide 1:1 | |
| dimethyl(dioctadecyl)azanium;bromide |
Specifications
| 3700-67-2 | |
| White | |
| Authentic | |
| Glass bottle | |
| (CH3)2N(Br)((CH2)17CH3)2 | |
| 5 g | |
| 1.047 | |
| Solubility in water: .. Other solubilities: soluble in acetonitrile,methanol,ethanol,aceton,e,ethyl acetate | |
| CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCCCC.[Br-] | |
| 630.96 | |
| 630.96 | |
| Granular Powder |
| 151.0°C to 157.0°C | |
| 1.0470g/mL | |
| 99.0 to 101% (Argentometry) | |
| C38H80BrN | |
| MFCD00041975 | |
| 04, IV, 829 | |
| dimethyldioctadecylammonium bromide, n,n-dimethyl-n-octadecyloctadecan-1-aminium bromide, dimethyldistearylammonium bromide, 1-octadecanaminium, n,n-dimethyl-n-octadecyl-, bromide, unii-z5t47r065a, dsab, distearyldimethylammonium bromide, dimethyldioctadecylazanium bromide, distearyl dimethyl ammonium bromide, 1-octadecanaminium, n,n-dimethyl-n-octadecyl-, bromide 1:1 | |
| PSLWZOIUBRXAQW-UHFFFAOYSA-M | |
| dimethyl(dioctadecyl)azanium;bromide | |
| 77293 | |
| 99+% | |
| Dimethyldioctadecylammonium bromide |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF
GHS Signal Word: Warning
EINECSNumber : 223-037-6
RUO – Research Use Only