missing translation for 'onlineSavingsMsg'
Learn More

Dimethylcarbamyl chloride, 96%

Catalog No. AAL0341522
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0341522 100 g
AAL0341514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0341522 Supplier Thermo Scientific Chemicals Supplier No. L0341522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 79-44-7 | C3H6ClNO | 107.537 g/mol

Dimethylcarbamyl chloride is used to make dyes and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylcarbamyl chloride is used to make dyes and pharmaceuticals.

Solubility
Reacts with water.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com

Chemical Identifiers

CAS 79-44-7
Molecular Formula C3H6ClNO
Molecular Weight (g/mol) 107.537
MDL Number MFCD00000635
InChI Key YIIMEMSDCNDGTB-UHFFFAOYSA-N
Synonym dimethylcarbamoyl chloride, dimethylcarbamyl chloride, dimethylcarbamic chloride, dimethylcarbamoylchloride, carbamic chloride, dimethyl, dimethylcarbamic acid chloride, dimethyl carbamoyl chloride, n,n-dimethylcarbamyl chloride, carbamoyl chloride, dimethyl, dimethylcarbamidoyl chloride
PubChem CID 6598
ChEBI CHEBI:82280
IUPAC Name N,N-dimethylcarbamoyl chloride
SMILES CN(C)C(=O)Cl

Specifications

Melting Point -33°C
Boiling Point 167°C to 168°C
Refractive Index 1.453
Linear Formula (CH3)2NCOCl
UN Number UN2262
Beilstein 878197
Solubility Information Reacts with water.
Density 1.171
Quantity 100 g
Flash Point 68°C (154°F)
Odor Unpleasant
Formula Weight 107.54
Sensitivity Moisture sensitive
Percent Purity 96%
Chemical Name or Material Dimethylcarbamyl chloride
Show More Show Less
Hazard Category H227-H302-H315-H319-H331-H335-H350
Hazard Statement GHS H Statement
H331-H350-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause cancer.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P235-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 201-208-6
RTECSNumber FD4200000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only