missing translation for 'onlineSavingsMsg'
Learn More

Dimethylcarbamoyl Chloride 98.0+%, TCI America™

Catalog No. p-7126379
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D069525G 25 g
D0695500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D069525G Supplier TCI America Supplier No. D069525G
Only null left

Chemical Identifiers

CAS 79-44-7
Molecular Formula C3H6ClNO
Molecular Weight (g/mol) 107.537
MDL Number MFCD00000635
InChI Key YIIMEMSDCNDGTB-UHFFFAOYSA-N
Synonym dimethylcarbamoyl chloride, dimethylcarbamyl chloride, dimethylcarbamic chloride, dimethylcarbamoylchloride, carbamic chloride, dimethyl, dimethylcarbamic acid chloride, dimethyl carbamoyl chloride, n,n-dimethylcarbamyl chloride, carbamoyl chloride, dimethyl, dimethylcarbamidoyl chloride
PubChem CID 6598
ChEBI CHEBI:82280
IUPAC Name N,N-dimethylcarbamoyl chloride
SMILES CN(C)C(=O)Cl

Specifications

Boiling Point 167°C
Color Yellow
UN Number 2262
Quantity 25 g
Formula Weight 107.54
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Dimethylcarbamoyl Chloride
EINECSNumber (2)-2858
RTECSNumber FD4200000
TSCA Yes