missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylbis(phenylethynyl)silane 98.0+%, TCI America™
$164.03 - $561.08
Chemical Identifiers
| CAS | 2170-08-3 |
|---|---|
| Molecular Formula | C18H16Si |
| Molecular Weight (g/mol) | 260.41 |
| MDL Number | MFCD00270967 |
| InChI Key | LRBLIVYQOCFXPX-UHFFFAOYSA-N |
| PubChem CID | 11010735 |
| IUPAC Name | dimethyl-bis(2-phenylethynyl)silane |
| SMILES | C[Si](C)(C#CC1=CC=CC=C1)C#CC2=CC=CC=C2 |
Chemical Identifiers
| 2170-08-3 | |
| 260.41 | |
| LRBLIVYQOCFXPX-UHFFFAOYSA-N | |
| dimethyl-bis(2-phenylethynyl)silane |
| C18H16Si | |
| MFCD00270967 | |
| 11010735 | |
| C[Si](C)(C#CC1=CC=CC=C1)C#CC2=CC=CC=C2 |
Specifications
| 2170-08-3 | |
| Yellow | |
| C18H16Si | |
| 1 g | |
| C[Si](C)(C#CC1=CC=CC=C1)C#CC2=CC=CC=C2 | |
| 260.41 | |
| 260.41 | |
| Crystalline Powder |
| 81°C | |
| 177°C | |
| MFCD00270967 | |
| LRBLIVYQOCFXPX-UHFFFAOYSA-N | |
| dimethyl-bis(2-phenylethynyl)silane | |
| 11010735 | |
| ≥98.0% (GC) | |
| Dimethylbis(phenylethynyl)silane |
Safety and Handling
TSCA : No