Learn More
Dimethylanilinium Tetrakis (pentafluorophenyl)borate, 98%
CAS: 118612-00-3 | C32H12BF20N | 801.23 g/mol
$2200.71 - $2200.71
Chemical Identifiers
| CAS | 118612-00-3 |
|---|---|
| Molecular Formula | C32H12BF20N |
| Molecular Weight (g/mol) | 801.23 |
| MDL Number | MFCD01074420 |
| InChI Key | BRHZQNMGSKUUMN-UHFFFAOYSA-O |
| Synonym | dimethylanilinium tetrakis pentafluorophenyl borate, unii-h8r86l92nd, n,n-dimethylanilinium tetrakis pentafluorophenyl borate, n,n-dimethylbenzenaminium tetrakis perfluorophenyl borate, n,n-dimethylanilinium tetra pentafluorophenyl borate, benzene, pentafluoro-, boron complex, n,n-dimethylanilinium tetrakis pentafluorophenyl borate 1-, benzenamine, n,n-dimethyl-, tetrakis pentafluorophenyl borate 1-, dimethyl phenyl ammonium tetrakis 2,3,4,5,6-pentafluorophenyl borate, borate 1-, tetrakis pentafluorophenyl-, hydrogen, compd. with n,n-dimethylbenzenamine 1:1:1 |
| PubChem CID | 10996402 |
| IUPAC Name | dimethyl(phenyl)azanium;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide |
| SMILES | [B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.C[NH+](C)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC382400050
|
Thermo Scientific Chemicals
382400050 |
5 g | Glass bottle |
Each for $2,200.71
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 118612-00-3 | |
| 801.23 | |
| BRHZQNMGSKUUMN-UHFFFAOYSA-O | |
| 10996402 | |
| [B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.C[NH+](C)C1=CC=CC=C1 |
| C32H12BF20N | |
| MFCD01074420 | |
| dimethylanilinium tetrakis pentafluorophenyl borate, unii-h8r86l92nd, n,n-dimethylanilinium tetrakis pentafluorophenyl borate, n,n-dimethylbenzenaminium tetrakis perfluorophenyl borate, n,n-dimethylanilinium tetra pentafluorophenyl borate, benzene, pentafluoro-, boron complex, n,n-dimethylanilinium tetrakis pentafluorophenyl borate 1-, benzenamine, n,n-dimethyl-, tetrakis pentafluorophenyl borate 1-, dimethyl phenyl ammonium tetrakis 2,3,4,5,6-pentafluorophenyl borate, borate 1-, tetrakis pentafluorophenyl-, hydrogen, compd. with n,n-dimethylbenzenamine 1:1:1 | |
| dimethyl(phenyl)azanium;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide |
Specifications
| 118612-00-3 | |
| Authentic | |
| Glass bottle | |
| MFCD01074420 | |
| dimethylanilinium tetrakis pentafluorophenyl borate, unii-h8r86l92nd, n,n-dimethylanilinium tetrakis pentafluorophenyl borate, n,n-dimethylbenzenaminium tetrakis perfluorophenyl borate, n,n-dimethylanilinium tetra pentafluorophenyl borate, benzene, pentafluoro-, boron complex, n,n-dimethylanilinium tetrakis pentafluorophenyl borate 1-, benzenamine, n,n-dimethyl-, tetrakis pentafluorophenyl borate 1-, dimethyl phenyl ammonium tetrakis 2,3,4,5,6-pentafluorophenyl borate, borate 1-, tetrakis pentafluorophenyl-, hydrogen, compd. with n,n-dimethylbenzenamine 1:1:1 | |
| [B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.C[NH+](C)C1=CC=CC=C1 | |
| 801.23 | |
| 801.23 | |
| Powder |
| Blue-White | |
| 98% | |
| C32H12BF20N | |
| 5 g | |
| BRHZQNMGSKUUMN-UHFFFAOYSA-O | |
| dimethyl(phenyl)azanium;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide | |
| 10996402 | |
| 98% | |
| Dimethylanilinium tetrakis (pentafluorophenyl)borate |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
Suspected of causing cancer.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical
GHS Signal Word: Danger
RUO – Research Use Only