missing translation for 'onlineSavingsMsg'
Learn More

Dimethylaminoacetaldehyde diethyl acetal, 95%, Thermo Scientific Chemicals

Catalog No. AAL0627214
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0627214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0627214 Supplier Thermo Scientific Chemicals Supplier No. L0627214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3616-56-6 | Molecular Formula: C8H19NO2 | Molecular Weight: 161.245 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3616-56-6
Molecular Formula C8H19NO2
Molecular Weight (g/mol) 161.245
MDL Number MFCD00009232
InChI Key SSFAUOAQOOISRQ-UHFFFAOYSA-N
Synonym dimethylamino acetaldehyde diethyl acetal, 2,2-diethoxy-n,n-dimethylethylamine, 2,2-diethoxyethyl dimethylamine, dimethylaminoacetaldehyde diethyl acetal, ethanamine, 2,2-diethoxy-n,n-dimethyl, dimethylaminoacetaldehyde diethylacetal, n-2,2-diethoxyethyl dimethylamine, 2,2-dieth, acmc-1ajh8
PubChem CID 77163
IUPAC Name 2,2-diethoxy-N,N-dimethylethanamine
SMILES CCOC(CN(C)C)OCC

Specifications

Density 0.876
Boiling Point 164°C to 166°C
Flash Point 45°C (113°F)
Odor Amine-like
Refractive Index 1.4115
Quantity 25 g
UN Number UN1993
Beilstein 1739149
Formula Weight 161.25
Percent Purity 95%
Chemical Name or Material Dimethylaminoacetaldehyde diethyl acetal
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 222-800-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only