missing translation for 'onlineSavingsMsg'
Learn More

(Dimethylamino)acetonitrile 96.0+%, TCI America™

Catalog No. D192925ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D192925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D192925ML Supplier TCI America Supplier No. D192925ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 926-64-7
Molecular Formula C4H8N2
Molecular Weight (g/mol) 84.12
MDL Number MFCD00001890
InChI Key PLXBWEPPAAQASG-UHFFFAOYSA-N
Synonym dimethylaminoacetonitrile, dimethylamino acetonitrile, acetonitrile, dimethylamino, 2-dimethylamino acetonitrile, n-cyanomethyl dimethylamine, glycinonitrile, n,n-dimethyl, unii-wu34f15n7c, glycinonitrile,n-dimethyl
PubChem CID 61237
IUPAC Name 2-(dimethylamino)acetonitrile
SMILES CN(C)CC#N

Specifications

Color Yellow
Boiling Point 138°C
Quantity 25 mL
UN Number 2378
Formula Weight 84.12
Percent Purity ≥96.0% (GC,T)
Grade Not Available
Physical Form Liquid
Chemical Name or Material (Dimethylamino)acetonitrile
RTECSNumber AL9450000
TSCA No