missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Dimethylamino)acetaldehyde Diethyl Acetal 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 60°C |
| Quantity | 25 mL |
| UN Number | 1993 |
| Formula Weight | 161.25 |
| Percent Purity | ≥97.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Liquid |
| Chemical Name or Material | (Dimethylamino)acetaldehyde Diethyl Acetal |
Safety and Handling
| EINECSNumber | (2)-0530 |
| TSCA | No |