missing translation for 'onlineSavingsMsg'
Learn More

Dimethylamine Hydroiodide 98.0+%, TCI America™

Catalog No. p-7128338
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45551G 1 g
D45555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45551G Supplier TCI America Supplier No. D45551G
Only null left

Chemical Identifiers

CAS 51066-74-1
Molecular Formula C2H8IN
Molecular Weight (g/mol) 173.00
MDL Number MFCD28100829
InChI Key JMXLWMIFDJCGBV-UHFFFAOYSA-N
Synonym Dimethylammonium Iodide
PubChem CID 12199010
IUPAC Name dimethylazanium iodide
SMILES [I-].C[NH2+]C

Specifications

Melting Point 155°C
Color White
Quantity 1 g
Formula Weight 173.00
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethylamine Hydroiodide