Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dimethylamine Hydroiodide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 155°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 173.00 |
| Percent Purity | ≥98.0% (N,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Dimethylamine Hydroiodide |
Safety and Handling
| TSCA | No |